Plasti-Peel® Low-Temperature Sterilization Pouches, Cardinal Health
Supplier: Cardinal
32115
CV32115CS 5298.5 USD
CV32115
Plasti-Peel® Low-Temperature Sterilization Pouches, Cardinal Health
Bags Sterilization Pouches
These clear film pouches are for all types of low-temperature sterilization.
  • Pouches feature an ETO/Plasma indicator and green print
They are made from a breathable, heavy-duty material for heat sensitive devices.