Acti-Lance® Push-Button Safety Lancets, HTL-Strefa
Supplier: HTL-STREFA
7156
10220-630BX 21.34 USD
10220-630
Acti-Lance® Push-Button Safety Lancets, HTL-Strefa
Acti-Lance® Safety Lancets efficiently provide an adequate blood sample with just a single push of the button
Ultra-sharp, electro-polished needles feature a silicone coating to minimize puncture force, reducing patient discomfort. The dual-spring assembly ensures a true linear needle path and rapid needle retraction, reducing tissue damage to promote rapid healing.
Acti-Lance® Safety Lancets are available in three formats. The Lite Safety Lancet is designed for the delicate skin of elderly and pediatric patients. The Universal Safety Lancet is designed for most common capillary blood tests. The Special Safety Lancet is bladed for use with larger sample sizes, patients with calloused skin, or as a heestick for neonates.
Gentle electro-polished, silicone-coated needles are linearly driven to assure lower pain and quick healing. Needles are supplied gamma-sterilized, fully covered, and protected with a sterility tab. An exclusive self-destructive activation mechanism eliminates the risk of misuse, or receiving an incorrect blood sample.
Certifications: Certified gamma sterilization.
Ordering information: Orders should be placed for case quantities.