Would you like to log in again?
Vizion TV/Monitor Stand and Mount for 35–55" TV, 80” Height, Marvel
Supplier: THE MARVEL GROUP, INC.
762805007690 762805007706 762805007652 762805007683 762805007331 762805007348 762805007676 762805007669
The Marvel Vizion Mobile Flat Panel Stand with a height adjustable bracket, built in cable management steel construction is ideal for commercial use! This 80” tall TV stand will easily hold a 35” - 55” Flat Panel TV
Built in cable management will help organize cables and wires. The mount (screen center) adjusts from 75.25” from floor to 77.25” from floor. This universal mount will attach to most Flat Panel TV’s weighing less than 200 lbs. Black finish. Powdercoat paint.
Models with the optional Microphone/Amplifier Module feature a 16 channel UHF wireless receiver. Frequency: 584 MHz - 608 MHz. Range: 300’ (91 m). 50 Watt Amplifier. Enclosure: Sealed type / Rotational molded. Mounting Bracket: 2U rackmount flanges. On/off switch. Line in volume control Microphone volume. Two 3.5 mm line in jacks. Rear. Speaker out - RCA. One 3.5 mm line in jack 12 DC power jack. XLR / 1/4” TRS (Tip Ring Sleeve) combo jack.
This attractive stand was designed for use in videoconferencing, mobile digital signage, classroom, and visual presentation environments. The heavy duty steel construction provides stability for high traffic areas, while the easy roll 4” swivel casters (four lockable) allow you to move the TV Stand where you need it.
ORDER
Expand All / Collapse All
Shelf Dimensions Includes Shelf Type Supplier No. VWR Catalog Number Unit Price Quantity
Unit Without Microphone/Amplifier 762805007331 10121-004 Each Retrieving
5W x 9"L, Camera Shelf Unit Without Microphone/Amplifier Unit with Camera Shelf 762805007348 10121-006 Each Retrieving
173/4"W x 14"L, Accessory Shelf Unit Without Microphone/Amplifier One Equipment Shelf 762805007652 10121-060 Each Retrieving
173/4"W x 14"L, Accessory Shelf Unit Without Microphone/Amplifier Two Equipment Shelves 762805007669 10121-062 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty