Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vizion Presenter Lockable Multimedia Cart with Four Side Shelves, Marvel

Supplier: THE MARVEL GROUP, INC.
762805005627 762805000806 762805000837 762805005597 762805005641 762805000813 762805005665 762805005610 762805005634 762805000820 762805005658 762805005603
The Vizion Presenter Multimedia Cart from Marvel is a steel cart with a high pressure laminate top designed for Multimedia presentations

High pressure laminate, powdercoat paint. Lockable doors; CPU, rack or component area.

Models with the optional Microphone/Amplifier module feature a 16 channel UHF wireless receiver. Frequency 584 MHz - 608 MHz. Range 300’ (91 m). 50 Watt Amplifier. Enclosure is Sealed type / Rotational molded. Mounting Bracket is 2U rackmount flanges. On/off switch. Line in volume control. Microphone volume. Two 3.5 mm line in jacks. Rear. Speaker out - RCA. One 3.5 mm line in jack. 12 DC power jack. XLR / 1/4” TRS (Tip Ring Sleeve) combo jack.

The cart features four shelves with retaining lip for collateral material as well as a top working surface for additional presentation items. Additionally, the cart features two locking storage drawers as well as pull-out front and side shelves-with detents. Cable management is handled via an interior wire management channel as well as a 2" grommet. The rear panel is readily accessible and vented to help with heat dissipation. Grilles are also included along with thread studs for mounting 4" speakers. There is also a shelf for 4" speakers (speakers sold separately).The lockable door includes two keys, and is infrared friendly to allow you to use a remote control even when the door is closed. There is sufficient space behind the door for a 6U rack piece of equipment. The entire cart is transported via four ball bearing casters using its side push handle.

Certifications: GREENGUARD Gold Certified.

Ordering information: Accessories/Props Not Included.



 

ORDER

Expand All / Collapse All
Door Type Finish Includes Supplier No. VWR Catalog Number Unit Price Quantity
Acrylic Door Cherry/Dark Neutral Without Microphone/Amplifier 762805000820 10120-812 Each Retrieving
Steel Door Oak/Dark Neutral Without Microphone/Amplifier 762805000813 10120-822 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping