Leishman's eosin-methylene blue solution, Sigma-Aldrich®

Synonyms: Eosin and methylene blue LeishmanEosin-Methylene blue dye LeishmanLeishman stainLeishman's stain (eosin methylene blue compound)Leishman's stain

R03087-76 R03087-76
EM-R03087-76EA 190 USD
EM-R03087-76 CAR03087-76
Leishman's eosin-methylene blue solution, Sigma-Aldrich®
Leishman's Eosin-Methylene blue

Leishman Stain Solution. Used for staining blood smears to differentiate blood corpuscles, malarial parasites, trypanosomers, etc. Can be used as an alternative to Giemsa.

MDL Number: MFCD00131498
CAS Number: 12627-53-1

Order Now

Specification Test Results

Absorbance at maximum (Azure) 0.30-0.40AU
Absorbance at maximum (Eosin) 0.22-0.30AU
Absorbance maximum wavelength (Azure) 645-655nm
Absorbance maximum wavelength (Eosin) 520-530nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR