Tetramethylammonium chloride

Supplier: MilliporeSigma

Synonyms: TMACl

8.45105.0250 8.45105.0250
EM8.45105.0250EA 82.02 USD InStock
EM8.45105.0250 CA8.45105.0250
Tetramethylammonium chloride
Tetramethylammonium chloride

Tetramethylammonium chloride Msynth®plus

Formula: (CH₃)₄NCl
MW: 109,6 g/mol
Melting Pt: Min. 300 °C
Density: 1.169 g/cm³ (20 °C)
Flash Pt: Min. 100 °C
Storage Temperature: ambient
MDL Number: MFCD00011628
CAS Number: 75-57-0
EINECS: 200-880-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Hill Formula C₄H₁₂ClN
Chemical formula (CH₃)₄NCl
Molar Mass 109.60 g/mol
Density 1.17 g/cm³ (20 °C)
pH value 6 - 8 (100 g/l, H₂O, 20 °C)
Vapor pressure (20 °C)
Solubility (20 °C) soluble
LD 50 oral LD50 Rat 50 mg/kg
LD 50 dermal LD50 Rat 537 mg/kg
Storage class 6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK WGK 1 slightly water endangering
R Phrase R 21-25
S Phrase S 36/37-45

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR