Dicyclopropyl ketone, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: DicyclopropylketoneDicyclopropylmethanone

M-Clarity™ quality level = MQ200

8.40157.0025 8.40157.0025
EM8.40157.0025CS 7580 USD
EM8.40157.0025 CA8.40157.0025
Dicyclopropyl ketone, Sigma-Aldrich®
Dicyclopropyl ketone

Dicyclopropylketone for synthesis

Formula: C₇H₁₀O
MW: 110.16 g/mol
MDL Number: MFCD00001276
CAS Number: 1121-37-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Hill Formula C₇H₁₀O
Molar Mass 110.15 g/mol
Boiling point 160 - 162 °C (1013 hPa)
Density 0.968 g/cm³
Flash point 39 °C
Storage class 3 Flammable liquids
WGK WGK 3 highly water endangering
R Phrase R 10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR