Caffeic acid, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
3,4-Dihydroxycinnamic acid
, 3-(3,4-Dihydroxyphenyl)acrylic acid
M-Clarity™ quality level = MQ200
8.22029.0010
8.22029.0010
EM8.22029.0010CS
6627
USD
CA8.22029.0010
EM8.22029.0010
Caffeic acid, Sigma-Aldrich®
Caffeic acid
3,4-Dihydroxycinnamic acid for synthesis
Formula:
C₉H₈O₄ MW: 180.16 g/mol |
MDL Number:
MFCD00004392 CAS Number: 331-39-5 |
Specification Test Results
Hill Formula | C₉H₈O₄ |
Chemical formula | (OH)₂C₆H₃CHCHCOOH |
Molar Mass | 180.15 g/mol |
Melting Point | 234 - 237 °C (decomposition) |
Solubility | (20 °C) slightly soluble |
Storage class | 10 - 13 Other liquids and solids |
WGK | WGK 3 highly water endangering |
R Phrase | R 40 |
S Phrase | S 36/37 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...