o-Ethyltoluene, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
1-Ethyl-2-methylbenzene
, 2-Ethyltoluene
M-Clarity™ quality level = MQ200
8.20044.0010
8.20044.0010
EM8.20044.0010CS
122474
USD
EM8.20044.0010
CA8.20044.0010
o-Ethyltoluene, Sigma-Aldrich®
o-Ethyltoluene
2-Ethyltoluene for synthesis
Formula:
C₉H₁₂ MW: 120.19 g/mol |
MDL Number:
MFCD00009257 CAS Number: 611-14-3 UN: 3295 ADR: 3,III |
Specification Test Results
Hill Formula | C₉H₁₂ |
Chemical formula | 2-(C₂H₅)C₆H₄CH₃ |
Molar Mass | 120.19 g/mol |
Boiling point | 164 - 165 °C (1013 hPa) |
Density | 0.88 g/cm³ (20 °C) |
Flash point | 53 °C |
Ignition temperature | 440 °C |
Melting Point | -17 °C |
Saturation concentration (air) | 14 g/m³ (20 °C) |
Vapor pressure | 2.8 hPa (20 °C) |
Refractive Index | 1.5039 (20 °C, 589 nm) |
Solubility | 74.6 mg/l (25 °C) (experimental) |
LD 50 oral | LD50 Rat 5000 mg/kg |
Storage class | 3 Flammable liquids |
WGK | WGK 2 water endangering |
R Phrase | R 10 |
S Phrase | S 24/25 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...