N-Methylformanilide, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.06169.0250 8.06169.0250
EM8.06169.0250CS 1061 USD
CA8.06169.0250 EM8.06169.0250
N-Methylformanilide, Sigma-Aldrich®
N-Methylformanilide

N-Methylformanilide for synthesis

Formula: C₈H₉NO
MDL Number: MFCD00003283
CAS Number: 93-61-8

Order Now

Specification Test Results

Hill Formula C₈H₉NO
Chemical formula HCON(CH₃)C₆H₅
Molar Mass 135.17 g/mol
Boiling point 243 - 244 °C (1013 hPa)
Density 1.095 g/cm³ (20 °C)
Flash point 126 °C
Melting Point 8 - 13 °C
pH value 3.5 - 5.5 (H₂O, 20 °C) (saturated aqueous solution)
Vapor pressure 2.66 Pa (25 °C)
Refractive Index 1.5593 (20 °C, 589 nm)
Solubility 10.3 g/l (25 °C)
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering
R Phrase R 38
S Phrase S 25

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR