Immersion Coolers, TC®, Huber
Supplier: Huber
3003.0046.00 3004.0022.00 3004.0006.99 3003.0008.99 3003.0006.99 3004.0008.99 3004.0021.00 3003.0045.00
10221-052EA 8437.05 USD
10221-052 10221-062 10221-050 10221-056 10221-054 10221-048 10221-058 10221-060
Immersion Coolers, TC®, Huber
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The TC range of immersion coolers are a flexible solution for a variaty of cooling tasks. They are universally used for rapid cooling of fluids.
  • Provide rapid cooling of fluids
  • Ideal for counter-cooling or as a replacement for dry ice
  • LED temperature display (E models)
  • Easy operation by means of only three keys
  • Compact, space-saving construction
  • High quality stainless steel Immersion probe
Typical applications include counter-cooling for heat circulation thermostats or replacing cooling by dry ice, they are also an environmentally friendly alternative to mains water cooling. The TC immersion coolers are suitable for unsupervised use, for cooling tasks which require continuous cooling or as a variant with temperature control and a PT100 probe. The models with suffix "E“ have an LED display with entry of nominal values and regulate with a precision of ±0,5 K. A spiral-shaped or flexible immersion cooling probe made out of stainless steel is available for all models so that fluids can be cooled in virtually all vessel types. The compact housing is made from high quality stainless steel, and is fitted with carrying grips. Units have a small footprint, require little installation area and optional accessory castors enable easy relocation around the laboratory.
Ordering information: Model with suffix 'F' available with flexible probes, contact your Avantor representative for further information.