Human Recombinant VAP-1 (from E. coli)
Supplier: Tonbo Biosciences
21-7090-U100 21-7090-U010 21-7090-U500 21-7090-U002
10274-056EA 226.91 USD
10274-056 10274-058 10002-600 10274-054
Human Recombinant VAP-1 (from E. coli)
Vascular adhesion protein­1 (VAP­1) is a type II membrane cell adhesion protein belonging to the copper/topaquinone oxidase family and functions both as an enzyme and an adhesion molecule for lymphocytes. VAP-1 is primarily expressed on the high endothelial venules of peripheral lymph nodes and on hepatic endothelia. Inhibition of VAP-1 can protect against inflammation related damage to certain injured tissues.
  • Carrier-free