Human Recombinant TIMP2 (from E. coli)
Supplier: Tonbo Biosciences
21-7086-U010 21-7086-U100 21-7086-U002
10273-540EA 98.28 USD
10273-540 10274-030 10002-428
Human Recombinant TIMP2 (from E. coli)
Tissue Inhibitors of Metalloproteinase-2 (TIMP-2) is one of four TIMP proteins that functions as an endogenous inhibitor for the matrix metalloproteinases (MMPs). An imbalance between MMPs and their respective TIMPs can lead to various disease states. TIMP-2 is widely expressed in many mammalian tissues. It participates in functions including wound healing, angiogenesis, inflammatory responses and tumor metastasis. TIMP-2 can also act through an MMP-independent mechanism to regulate cell proliferation and demonstrates anti-angiogenic activities in vivo.
  • Carrier-free