Human Recombinant TIMP1 (from E. coli)
Supplier: Tonbo Biosciences
21-7085-U100 21-7085-U500 21-7085-U010 21-7085-U002
10002-426EA 1240.01 USD
10002-426 10273-538 10273-536 10273-534
Human Recombinant TIMP1 (from E. coli)
Tissue Inhibitors of Metalloproteinase-1 (TIMP-1) is one of four TIMP proteins that functions as an endogenous inhibitor for the matrix metalloproteinases (MMPs). An imbalance between MMPs and their respective TIMPs can lead to various disease states. TIMP-1 is widely expressed in many mammalian tissues, notably in the reproductive organs. It has a wide variety of roles, participating in functions including wound healing, angiogenesis, inflammatory responses and tumor metastasis. TIMP-1 appears to be the only of the four proteins that inhibit some MMPs less well than others – it is a poor inhibitor of MMP-19 and several MT-MMPs.
  • Carrier-free