Human Recombinant NT4 (from E. coli)
Supplier: Tonbo Biosciences
21-7078-U500 21-7078-U010 21-7078-U100 21-7078-U002
10002-412EA 1343.96 USD
10002-412 10273-970 10273-968 10273-966
Human Recombinant NT4 (from E. coli)
NT-4 (Neurotrophin-4) is a member of the NGF family of neurotrophins that is comprised of at least four proteins including NGF, BDNF, NT-3, and NT-4/5. Neurotrophins are critical to the development of the central and peripheral nervous system and they play important roles in injury induced neuron regeneration. NT-3 is expressed in the prostate, thymus, placenta and skeletal muscle and promotes the survival of peripheral sensory sympathetic neurons. NT-4 is a secreted protein that primarily signals through the LNGFR and trkB receptors.
  • Carrier-free