Mouse Recombinant Noggin (from E. coli)
Supplier: Tonbo Biosciences
21-8004-U100 21-8004-U005 21-8004-U020 21-8004-U500
10273-950EA 226.91 USD
10273-950 10002-612 10273-948 10273-952
Mouse Recombinant Noggin (from E. coli)
Noggin is a highly conserved molecule and is a secreted protein that drives neural formation and dorsalization during embryonic development. It is also a BMP antagonist, biding BMP-4 as well as BMP-2 and BMP-7, to prevent them from associating with their receptors. Noggin is expressed in defined areas of the adult central nervous system and in various peripheral tissues such as lung, skeletal muscle, and skin.
  • Carrier-free