Mouse Recombinant SCD40 Ligand (from E. coli)
Supplier: Tonbo Biosciences
21-8512-U005 21-8512-U100 21-8512-U025 21-8512-U500
10002-148EA 811.53 USD
10002-148 10273-510 10273-514 10273-512
Mouse Recombinant SCD40 Ligand (from E. coli)
CD40 ligand, also known as CD154, is a cell surface glycoprotein expressed on the surface of activated T cells. Binding of CD40 ligand to CD40, expressed on B cells, stimulates B cell proliferation and secretion of all immunoglobulin isotypes.
  • Carrier-free