Mouse Recombinant CCL7 (from E. coli)
Supplier: Tonbo Biosciences
21-7011-U100 21-7011-U002 21-7011-U500 21-7011-U010
10273-632EA 1931.9 USD
10273-632 10273-630 10002-140 10273-628
Mouse Recombinant CCL7 (from E. coli)
CCL7, also known as MCP3, is a member of the CC family of chemokines. CCL7 is a chemoattractant for monocytes and eosinophils and can regulate the functional activity of monocytes. CCL7 exerts its effects through binding to the receptors CCR1, CCR2 and CCR3.
  • Carrier-free