Dinickel boride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Nickel boride

M-Clarity™ quality level = MQ200

8.43836.0010 8.43836.0010
EM8.43836.0010CS 118718.45 USD
CA8.43836.0010 EM8.43836.0010
Dinickel boride for synthesis, Sigma-Aldrich®
Dinickel boride
Formula: BNi₂
MDL Number: MFCD00084757
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Molar Mass 128.2 g/mol
Hill Formula BNi2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR