Hexachloroplatinic(IV) acid 25 - 50% in isopropanol (IPA) (11-15% Pt) for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.43812.0005 8.43812.0005 8.43812.0001 8.43812.0001
EM8.43812.0005CS 40272.11 USD
EM8.43812.0005 EM8.43812.0001 CA8.43812.0005 CA8.43812.0001
Hexachloroplatinic(IV) acid 25 - 50% in isopropanol (IPA) (11-15% Pt) for synthesis, Sigma-Aldrich®
Hexachloroplatinic (IV) acid
Formula: H₂Cl₆Pt
MW: 409.81 g/mol
Boiling Pt: 82.4 °C (1013 hPa) (2…Propanol)
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011330
CAS Number: 16941-12-1
UN: 3286
ADR: 3 (6.1, 8),II

Order Now

Specification Test Results

Assay (of Pt) 11.0 - 15.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR