4-Nitrophenyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1-Acetoxy-4-nitrobenzene

M-Clarity™ quality level = MQ200

8.43075.0025 8.43075.0025
EM8.43075.0025CS 13114 USD
EM8.43075.0025 CA8.43075.0025
4-Nitrophenyl acetate for synthesis, Sigma-Aldrich®
4-Nitrophenyl acetate
Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD00007326
CAS Number: 830-03-5

Order Now

Specification Test Results

Solubility 0.53 g/l (25 °C)
Melting Point 77 - 79 °C
Molar Mass 181.15 g/mol
Flash Point 110 °C
Hill Formula C8H7NO4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR