1,4-Bis(2-methylstyryl)benzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Bis-MSB1-Methyl-2-(2-(4-[2-(2-methylphenyl)ethenyl]phenyl)ethenyl)benzene1,4-Di(2-methylstyryl)benzenep-Bis(o-Methylstyryl) Benzene

M-Clarity™ quality level = MQ200

8.41875.0001 8.41875.0001
EM8.41875.0001CS 26467 USD
EM8.41875.0001 CA8.41875.0001
1,4-Bis(2-methylstyryl)benzene for synthesis, Sigma-Aldrich®
1,4-Bis(2-methylstyryl)benzene
Formula: C₂₄H₂₂
MW: 310.44 g/mol
MDL Number: MFCD00008529
CAS Number: 13280-61-0

Order Now

Specification Test Results

Melting Point 179 - 181 °C
Molar Mass 310.43 g/mol
Synonyms Bis-MSB
Hill Formula C24H22

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR