4,4'-Anisoin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Anisoin2-Hydroxy-1,2-bis(4-methoxyphenyl)ethanone

M-Clarity™ quality level = MQ200

8.41595.0025 8.41595.0025
EM8.41595.0025CS 8100 USD
EM8.41595.0025 CA8.41595.0025
4,4'-Anisoin for synthesis, Sigma-Aldrich®
4,4'-Anisoin
Formula: C₁₆H₁₆O₄
MW: 272.30 g/mol
MDL Number: MFCD00008411
CAS Number: 119-52-8

Order Now

Specification Test Results

Melting Point 111 - 112 °C
Molar Mass 272.3 g/mol
Synonyms 4,4'-Dimethoxybenzoin
Hill Formula C16H16O4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR