1,3,5-Triisopropylbenzene for synthesis, Sigma-Aldrich®


M-Clarity™ quality level = MQ200

8.41587.0050 8.41587.0050
EM8.41587.0050CS 14806.27 USD
CA8.41587.0050 EM8.41587.0050
1,3,5-Triisopropylbenzene for synthesis, Sigma-Aldrich®
1,3,5-Triisopropylbenzene
Formula: C₁₅H₂₄
MW: 204.36 g/mol
MDL Number: MFCD00008890
CAS Number: 717-74-8

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -7 °C
Molar Mass 204.35 g/mol
Boiling Point 235 - 237 °C
Flash Point 86 °C
Density 0.85 g/cm3 (20 °C)
Hill Formula C15H24

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR