2-Bromoterephthalic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.41467.0005 8.41467.0025 8.41467.0025
EM8.41467.0025CS 37638 USD
CA8.41467.0005 CA8.41467.0025 EM8.41467.0025
2-Bromoterephthalic acid for synthesis, Sigma-Aldrich®
2-Bromoterephthalic acid
Formula: C₈H₅BrO₄
MDL Number: MFCD00002403
CAS Number: 586-35-6

Order Now

Specification Test Results

Melting Point 300 °C
Molar Mass 245.02 g/mol
Synonyms 2-Bromo-1,4-benzenedicarboxylic acid
Hill Formula C8H5BrO4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR