α-Benzoinoxime for synthesis, Sigma-Aldrich®
Synonyms:
α-Benzoin oxime
, 2-Hydroxy-1,2-diphenylethanone oxime
M-Clarity™ quality level = MQ200
8.41378.0025
8.41378.0100
8.41378.0100
8.41378.0025
EM8.41378.0025CS
7593.95
USD
EM8.41378.0025
CA8.41378.0100
CA8.41378.0025
EM8.41378.0100
α-Benzoinoxime for synthesis, Sigma-Aldrich®
α-Benzoinoxime
Formula:
C₁₄H₁₃NO₂ MW: 227.26 g/mol |
MDL Number:
MFCD00004501 CAS Number: 441-38-3 |
Specification Test Results
Melting Point | 153 - 154 °C |
Molar Mass | 227.26 g/mol |
Synonyms | Cuprone |
Hill Formula | C14H13NO2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...