Tetramethylammonium acetate 50% in water for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.41249.0010 8.41249.0010
EM8.41249.0010CS 173400.23 USD
EM8.41249.0010 CA8.41249.0010
Tetramethylammonium acetate 50% in water for synthesis, Sigma-Aldrich®
Tetramethylammonium acetate

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.000 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR