Sodium 1-hexadecanesulfonate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Sodium hexadecane-1-sulphonate
, Hexadecane-1-sulphonic acid sodium salt
, Hexadecane-1-sulfonic acid sodium salt
, Sodium hexadecane-1-sulfonate
M-Clarity™ quality level = MQ200
8.41813.0010
8.41813.0010
EM8.41813.0010CS
99528
USD
CA8.41813.0010
EM8.41813.0010
Sodium 1-hexadecanesulfonate for synthesis, Sigma-Aldrich®
Sodium 1-hexadecanesulphonate
Specification Test Results
Melting Point | 265.3 °C |
Molar Mass | 328.48 g/mol |
Hill Formula | C16H33NaO3S |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...