(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
ONPG
, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
, o-Nitrophenyl-β-D-galactoside
, O-Nitrophenyl-Β-D-Galactopyranoside
M-Clarity™ quality level = MQ200
8.41747.0001
8.41747.0001
EM8.41747.0001CS
39215
USD
CA8.41747.0001
EM8.41747.0001
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) for synthesis, Sigma-Aldrich®
(2-Nitrophenyl) β-D-galactopyranoside
Formula:
C₁₂H₁₅NO₈ MW: 301.25 g/mol |
MDL Number:
MFCD00063255 CAS Number: 369-07-3 |
Specification Test Results
Molar Mass | 301.25 g/mol |
Hill Formula | C12H15NO8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...