(2-Naphthoxy)acetic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Naphthoxyacetic acid

M-Clarity™ quality level = MQ200

8.41023.0100 8.41023.0100
EM8.41023.0100CS 924 USD
CA8.41023.0100 EM8.41023.0100
(2-Naphthoxy)acetic acid for synthesis, Sigma-Aldrich®
(2-Naphthoxy)acetic acid
Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
MDL Number: MFCD00004066
CAS Number: 120-23-0

Order Now

Specification Test Results

Melting Point 155 - 157 °C
Molar Mass 202.21 g/mol
Hill Formula C12H10O3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR