Ethyl salicylate for synthesis, Sigma-Aldrich®

Warning

Synonyms: Ethyl 2-hydroxybenzoate

M-Clarity™ quality level = MQ200

8.41018.1000 8.41018.1000 8.41018.0250
EM8.41018.1000CS 770.01 USD
EM8.41018.1000 CA8.41018.0250 CA8.41018.1000
Ethyl salicylate for synthesis, Sigma-Aldrich®
Ethyl salicylate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00002215
CAS Number: 118-61-6

Order Now

Specification Test Results

Solubility 0.25 g/l (20 °C)
Melting Point 1.3 °C
Molar Mass 166.17 g/mol
Boiling Point 232 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 100 °C
Explosion Limit 1.1 %(V)
Density 1.13 g/cm3 (20 °C)
Synonyms Salicylic acid ethyl ester, Ethyl-2-hydroxybenzoate
Hill Formula C9H10O3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR