4-(2-Pyridylazo)resorcinol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4-(2'-Pyridylazo)resorcinol

M-Clarity™ quality level = MQ200

8.41016.0005 8.41016.0005
EM8.41016.0005CS 114030 USD
CA8.41016.0005 EM8.41016.0005
4-(2-Pyridylazo)resorcinol for synthesis, Sigma-Aldrich®
4-(2-Pyridylazo)resorcinol

Order Now

Specification Test Results

Melting Point 187 - 189 °C
Molar Mass 215.21 g/mol
Synonyms 4-(2-Pyridinylazo)-1,3-dihydroxybenzene
Hill Formula C11H9N3O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR