4-Nitro-m-phenylenediamine moistened for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 4-Nitro-1,3-phenylenediamine

M-Clarity™ quality level = MQ200

8.40107.0025 8.40107.0025
EM8.40107.0025CS 7273 USD
EM8.40107.0025 CA8.40107.0025
4-Nitro-m-phenylenediamine moistened for synthesis, Sigma-Aldrich®
4-Nitro-m-phenylenediamine
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
MDL Number: MFCD00025289
CAS Number: 5131-58-8

Order Now

Specification Test Results

Solubility 1 g/l (15 °C)
Melting Point 153 °C (decomposition)
Bulk Density 550 kg/m3
Synonyms 1,3-Diamino-4-nitrobenzene

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR