(-)-trans-2-tert-Butylcyclohexanol for synthesis, Sigma-Aldrich®
M-Clarity™ quality level = MQ200
8.24509.0500
8.24509.0500
EM8.24509.0500CS
9702.22
USD
CA8.24509.0500
EM8.24509.0500
(-)-trans-2-tert-Butylcyclohexanol for synthesis, Sigma-Aldrich®
(-)-trans-2-tert-Butylcyclohexanol
Formula:
C₁₀H₂₀O |
CAS Number:
98104-30-4 |
Specification Test Results
Molar Mass | 156.27 g/mol |
Hill Formula | C10H20O |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...