(+)-trans-2-tert-Butylcyclohexanol for synthesis, Sigma-Aldrich®
M-Clarity™ quality level = MQ200
8.24508.0500
8.24508.0500
EM8.24508.0500CS
10400.54
USD
CA8.24508.0500
EM8.24508.0500
(+)-trans-2-tert-Butylcyclohexanol for synthesis, Sigma-Aldrich®
(+)-trans-2-tert-Butylcyclohexanol
Formula:
C₁₀H₂₀O |
CAS Number:
13492-07-4 |
Specification Test Results
Melting Point | 54 - 57 °C |
Molar Mass | 156.26 g/mol |
Boiling Point | 88 °C (10.7 hPa) |
Hill Formula | C10H20O |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...