(-)-Shikimic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
(3R,4S,5R)-3,4,5-Trihydroxycyclohex-1-ene-1-carboxylic acid
M-Clarity™ quality level = MQ200
8.24476.1000
8.24476.1000
8.24476.0250
8.24476.0250
EM8.24476.1000CS
13614
USD
EM8.24476.1000
EM8.24476.0250
CA8.24476.0250
CA8.24476.1000
(-)-Shikimic acid for synthesis, Sigma-Aldrich®
(-)-Shikimic acid
Formula:
C₇H₁₀O₅ MW: 174.15 g/mol |
MDL Number:
MFCD00066278 CAS Number: 138-59-0 |
Specification Test Results
Solubility | 180 g/l (20 °C) |
Melting Point | 190 °C |
Molar Mass | 174.15 g/mol |
Synonyms | (3R,4S,5R)-3,4,5-Trihydroxy-1-cyclohexenecarboxylic acid |
Hill Formula | C7H10O5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...