6-Azauracil for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1,2,4-Triazine-3,5(2H,4H)-dione

M-Clarity™ quality level = MQ200

8.41156.0025 8.41156.0005 8.41156.0025 8.41156.0005
EM8.41156.0025CS 44930 USD
CA8.41156.0025 CA8.41156.0005 EM8.41156.0025 EM8.41156.0005
6-Azauracil for synthesis, Sigma-Aldrich®
6-Azauracil
Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
MDL Number: MFCD00006456
CAS Number: 461-89-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting Point 274 - 275 °C
Molar Mass 113.07 g/mol
Synonyms 1,2,4-Triazine-3,5(2H,4H)-dione
Hill Formula C3H3N3O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR