3,4-Dichlorobenzyl alcohol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (3,4-Dichlorophenyl)methanol

M-Clarity™ quality level = MQ200

8.41133.0010 8.41133.0010
EM8.41133.0010CS 5544 USD
EM8.41133.0010 CA8.41133.0010
3,4-Dichlorobenzyl alcohol for synthesis, Sigma-Aldrich®
3,4-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
MDL Number: MFCD00004633
CAS Number: 1805-32-9
ADR: 6.1,III

Order Now

Specification Test Results

Melting Point 38 - 39 °C
Molar Mass 177.03 g/mol
Boiling Point 148 - 151 °C
Flash Point 110 °C
Synonyms 3,4-Dichloro-1-(hydroxymethyl)-benzene
Hill Formula C7H6Cl2O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR