1-Chlorooctadecane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: n-Octadecylchloride

M-Clarity™ quality level = MQ200

8.22000.0100 8.22000.0100
EM8.22000.0100CS 959 USD
CA8.22000.0100 EM8.22000.0100
1-Chlorooctadecane for synthesis, Sigma-Aldrich®
1-Chlorooctadecane
Formula: C₁₈H₃₇Cl
MW: 288.94 g/mol
MDL Number: MFCD00000960
CAS Number: 3386-33-2

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 20 - 23 °C
Molar Mass 288.94 g/mol
Boiling Point 165 - 166 °C (3 hPa)
Vapor Pressure 0.25 hPa (137 °C)
Flash Point 184 °C
Refractive Index 1.452 (20 °C, 589 nm)
Density 0.86 g/cm3 (20 °C)
Ignition Point 320 °C
Synonyms Stearyl chloride, Octadecyl chloride
Hill Formula C18H37Cl
Chemical Formula CH3(CH2)17Cl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR