4-Formylbenzoic acid for synthesis, halal, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Terephthalaldehydic acid4-Carboxybenzaldehyde

M-Clarity™ quality level = MQ200

8.21075.0010
EM8.21075.0010CS 7051 USD
EM8.21075.0010
4-Formylbenzoic acid for synthesis, halal, Sigma-Aldrich®
4-Formylbenzoic acid
Formula: C₈H₆O₃
MW: 150.13 g/mol
MDL Number: MFCD00006951
CAS Number: 619-66-9

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 248 - 250 °C
Molar Mass 150.14 g/mol
Bulk Density 396 kg/m3
pH 3.5 (1 g/l, H2O, 20 °C)
Synonyms 4-Formylbenzoic acid, 4-Carboxybenzaldehyde
Hill Formula C8H6O3
Chemical Formula 4-(OHC)C6H4COOH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR