Phenothiazine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
10H-Phenothiazine
M-Clarity™ quality level = MQ200
8.20975.1000
8.20975.1000
8.20975.0100
8.20975.0100
EM8.20975.1000CS
414
USD
CA8.20975.0100
CA8.20975.1000
EM8.20975.1000
EM8.20975.0100
Phenothiazine for synthesis, Sigma-Aldrich®
Phenothiazine
Formula:
C₁₂H₉NS MW: 199.28 g/mol |
MDL Number:
MFCD00005015 CAS Number: 92-84-2 UN: 3077 ADR: 9,III |
Specification Test Results
Solubility |
|
Melting Point | 182 - 185 °C |
Molar Mass | 199.26 g/mol |
Bulk Density | 650 kg/m3 |
Boiling Point | 381 °C (1011 hPa) |
Vapor Pressure |
|
Density | 1.34 g/cm3 (20 °C) |
pH | 6 (10 g/l, H2O, 20 °C) (aqueous suspension) |
Ignition Point | 470 °C |
Hill Formula | C12H9NS |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...