2-Naphthyl phenyl ketone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-Benzoylnaphthalene

M-Clarity™ quality level = MQ200

8.20869.0001 8.20869.0001
EM8.20869.0001CS 233579 USD
CA8.20869.0001 EM8.20869.0001
2-Naphthyl phenyl ketone for synthesis, Sigma-Aldrich®
2-Naphthyl phenyl ketone
Formula: C₁₇H₁₂O
MW: 232.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004106
CAS Number: 644-13-3

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 80 - 82 °C
Molar Mass 232.28 g/mol
Boiling Point 398 °C
Synonyms Phenyl 2-naphthyl ketone
Hill Formula C17H12O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR