1-Methyl-4-piperidone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
N-Methyl-4-piperidone
M-Clarity™ quality level = MQ200
8.20828.0100
8.20828.0100
EM8.20828.0100CS
2422
USD
EM8.20828.0100
CA8.20828.0100
1-Methyl-4-piperidone for synthesis, Sigma-Aldrich®
1-Methyl-4-piperidone
Formula:
C₆H₁₁NO MW: 113.16 g/mol |
MDL Number:
MFCD00006191 CAS Number: 1445-73-4 UN: 1993 ADR: 3,III |
Specification Test Results
Solubility | (20 °C) soluble |
Molar Mass | 113.16 g/mol |
Boiling Point | 60 - 62 °C (19 hPa) |
Flash Point | 58 °C |
Refractive Index | 1.4605 (20 °C, 589 nm) |
Density | 0.97 g/cm3 (20 °C) |
pH | 12 (100 g/l, H2O, 20 °C) |
Synonyms | N-Methyl-γ-piperidone |
Hill Formula | C6H11NO |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...