Benzimidazole for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
1H-Benzimidazole
, 1H-Benzo[d]imidazole
M-Clarity™ quality level = MQ200
8.21956.0100
8.21956.0100
8.21956.0500
8.21956.0500
EM8.21956.0500CS
1605
USD
EM8.21956.0500
EM8.21956.0100
CA8.21956.0500
CA8.21956.0100
Benzimidazole for synthesis, Sigma-Aldrich®
Benzimidazole
Formula:
C₇H₆N₂ MW: 118.14 g/mol |
MDL Number:
MFCD00005585 CAS Number: 51-17-2 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 170 - 173 °C |
Molar Mass | 118.13 g/mol |
Boiling Point |
|
Hill Formula | C7H6N2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...