1-Ethylquinaldinium iodide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1-Ethyl-2-methylquinolin-1-ium iodide1-Ethyl-2-methylquinolinium iodide

M-Clarity™ quality level = MQ200

8.21935.0050 8.21935.0050
EM8.21935.0050CS 2653 USD
CA8.21935.0050 EM8.21935.0050
1-Ethylquinaldinium iodide for synthesis, Sigma-Aldrich®
1-Ethylquinaldinium iodide
Formula: C₁₂H₁₄IN
MW: 299.15 g/mol
MDL Number: MFCD00031829
CAS Number: 606-55-3

Order Now

Specification Test Results

Solubility 250 g/l (20 °C)
Molar Mass 299.15 g/mol
Synonyms N-Ethyl-2-methylquinolinium iodide
Hill Formula C12H14IN

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR