N,N-Diethylformamide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Formdiethylamide
, Formic acid diethylamide
M-Clarity™ quality level = MQ200
8.21752.0100
8.21752.0100
8.21752.0250
8.21752.0250
EM8.21752.0250CS
2571
USD
EM8.21752.0250
CA8.21752.0250
EM8.21752.0100
CA8.21752.0100
N,N-Diethylformamide for synthesis, Sigma-Aldrich®
N,N-Diethylformamide
Formula:
C₅H₁₁NO MW: 101.15 g/mol |
MDL Number:
MFCD00003287 CAS Number: 617-84-5 |
Specification Test Results
Solubility | 1000 g/l (20 °C) |
Molar Mass | 101.15 g/mol |
Boiling Point | 178 °C |
Flash Point | 65 °C |
Refractive Index | 1.434 (20 °C, 589 nm) |
Density | 0.91 g/cm3 (20 °C) |
Synonyms | Formic acid diethylamide |
Hill Formula | C5H11NO |
Chemical Formula | HCON(C2H5)2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...