N-(Hydroxymethyl)acrylamide 48% in aqueous solution stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N-Methylolacrylamide

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.20675.1000 8.20675.1000
EM8.20675.1000CS 477 USD
EM8.20675.1000 CA8.20675.1000
N-(Hydroxymethyl)acrylamide 48% in aqueous solution stabilized for synthesis, Sigma-Aldrich®
N-(Hydroxymethyl)acrylamide
Formula: C₄H₇NO₂
MW: 101.11 g/mol
MDL Number: MFCD00004597
CAS Number: 924-42-5

Order Now

Specification Test Results

Solubility 1880 g/l (20 °C)
Melting Point -10 °C
Boiling Point 100 °C (1013 hPa)
Vapor Pressure 31 hPa (25 °C)
Flash Point 93 °C
Density 1.08 g/cm3 (20 °C)
pH 6.0 - 7.0 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR