Fluorene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Diphenylenmethane
, 9H-Fluorene
8.20572.0100
EM8.20572.0100CS
1505
USD
EM8.20572.0100
Fluorene for synthesis, Sigma-Aldrich®
Fluorene
Formula:
C₁₃H₁₀ MW: 166.22 g/mol |
MDL Number:
MFCD00001111 CAS Number: 86-73-7 UN: 3077 ADR: 9,III |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 113 - 115 °C |
Molar Mass | 166.22 g/mol |
Boiling Point | 295 °C |
Vapor Pressure | 13 hPa (146 °C) |
Flash Point | 152 °C |
Synonyms | Diphenylenemethane |
Hill Formula | C13H10 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...