N,N'-Diphenylbenzidine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: N4,N4'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine4,4'-Bis(phenylamino)biphenyl

M-Clarity™ quality level = MQ200

8.20530.0010 8.20530.0010
EM8.20530.0010CS 11445 USD
EM8.20530.0010 CA8.20530.0010
N,N'-Diphenylbenzidine for synthesis, Sigma-Aldrich®
N,N'-Diphenylbenzidine
Formula: C₂₄H₂₀N₂
MW: 336.44 g/mol
MDL Number: MFCD00003016
CAS Number: 531-91-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 245 - 250 °C
Molar Mass 336.43 g/mol
Synonyms N,N'-Diphenyl-4,4'-diaminobiphenyl, N,N'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine
Hill Formula C24H20N2
Chemical Formula 4-(C6H5NH)C6H4C6H4-4-(NHC6H5)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR