N,N-Diphenylacetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: N-Acetyldiphenylamine

M-Clarity™ quality level = MQ200

8.20524.0025 8.20524.0025
EM8.20524.0025CS 9171 USD
EM8.20524.0025 CA8.20524.0025
N,N-Diphenylacetamide for synthesis, Sigma-Aldrich®
N,N-Diphenylacetamide
Formula: C₁₄H₁₃NO
MDL Number: MFCD00008685
CAS Number: 519-87-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 98 - 101 °C
Molar Mass 211.26 g/mol
Synonyms Acetic acid diphenylamide
Hill Formula C14H13NO
Chemical Formula CH3CON(C6H5)2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR